Assay Details
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elp5 | NM_018740.2 | 2 | 85 | NP_061210.2 |
| AF004107.1 | AAC16252.1 | |||
| AK152582.1 | 2 | 89 | BAE31332.1 | |
| AK217337.1 | 2 | 102 |

Genomic Map