Assay Details
Target Gene Details
Entrez Gene ID: | 27364 |
Gene Name: | serine racemase |
Gene Aliases: |
Srs |
Location: |
Chr.11:74905047-74925798 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srr | NM_001163311.1 | 8 | 2648 | NP_001156783.1 |
| NM_013761.4 | 9 | 2874 | NP_038789.1 | |
| AK031687.1 | 8 | 2671 | BAC27514.1 | |
| AK043738.1 | 9 | 2780 | BAC31637.1 | |
| AK079343.1 | 5 | 2158 | ||
| AK157122.1 | 8 | 2799 | BAE33968.1 |
Target Gene Details
Entrez Gene ID: | 104662 |
Gene Name: | TSR1 20S rRNA accumulation |
Gene Aliases: |
AU040765, AW550801, mKIAA1401 |
Location: |
Chr.11:74898080-74909340 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsr1 | NM_177325.3 | NP_796299.2 | ||
| AK129350.1 | BAC98160.1 | |||
| AK154063.1 | BAE32349.1 | |||
| AK162686.1 | BAE37023.1 |
More Information
Set Membership: |
|

Genomic Map