Assay Details
Target Gene Details
Entrez Gene ID: | 237860 |
Gene Name: | slingshot homolog 2 (Drosophila) |
Gene Aliases: |
SSH-2, SSH-2L, mSSH-2L |
Location: |
Chr.11:77216169-77460220 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssh2 | NM_001291190.1 | 14 | 3787 | NP_001278119.1 |
| NM_177710.4 | 15 | 3651 | NP_808378.2 | |
| AB099288.1 | 15 | 3651 | BAC97811.1 | |
| AK155232.1 | 14 | 3778 | BAE33137.1 | |
| AK173243.1 | 4 | 2515 | BAD32521.1 | |
| BC141392.1 | 15 | 3645 | ||
| BC141393.1 | 15 | 3645 |
More Information
Set Membership: |
|

Genomic Map