Assay Details
Target Gene Details
Entrez Gene ID: | 21973 |
Gene Name: | topoisomerase (DNA) II alpha |
Gene Aliases: |
Top-2 |
Location: |
Chr.11:98992947-99024206 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top2a | NM_011623.2 | 36 | 4834 | NP_035753.2 |
| AK138079.1 | 10 | 1266 | BAE23548.1 | |
| BC094326.1 | 10 | 1587 | AAH94326.1 | |
| U01915.1 | 8 | 954 | AAC52135.1 |
More Information
Set Membership: |
|

Genomic Map