Assay Details
Target Gene Details
Entrez Gene ID: | 27225 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 |
Gene Aliases: |
1700055J08Rik, 2510027P10Rik, AI649272 |
Location: |
Chr.12:103407976-103425867 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx24 | NM_001159502.1 | 2 | 624 | NP_001152974.1 |
| NM_020494.3 | 2 | 458 | NP_065240.2 | |
| AF214732.1 | 2 | 438 | AAG02170.1 | |
| AK011012.1 | 2 | 402 | BAB27330.1 | |
| AK018862.1 | 2 | 440 | ||
| AK036538.1 | 2 | 422 | ||
| AK134339.1 | 2 | 436 | BAE22104.1 | |
| AK149810.1 | 2 | 409 | ||
| AK150778.1 | 2 | 410 | BAE29843.1 | |
| AK152551.1 | 2 | 410 | BAE31305.1 | |
| AK152928.1 | 2 | 410 | BAE31603.1 | |
| AK166360.1 | 2 | 416 | BAE38731.1 | |
| AK172557.1 | 2 | 409 | ||
| BC055048.1 | 2 | 388 | AAH55048.1 | |
| BC055317.1 | 2 | 624 | AAH55317.1 | |
| CJ141277.1 | 2 | 458 |

Genomic Map