Assay Details
Target Gene Details
Entrez Gene ID: | 28114 |
Gene Name: | NOL1/NOP2/Sun domain family member 2 |
Gene Aliases: |
D13Wsu123e, Misu |
Location: |
Chr.13:69612016-69635780 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun2 | NM_145354.5 | 19 | 3810 | NP_663329.3 |
| AK030124.1 | 18 | 3695 | BAC26795.1 |
More Information
Set Membership: |
|

Genomic Map