Assay Details
Target Gene Details
Entrez Gene ID: | 217734 |
Gene Name: | protein-O-mannosyltransferase 2 |
Gene Aliases: |
A830009D15Rik, AW046274 |
Location: |
Chr.12:87106861-87147902 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt2 | NM_153415.3 | 21 | 4654 | NP_700464.2 |
| AK089079.1 | 1 | 149 | ||
| AK211754.1 | ||||
| AY090481.1 | 21 | 4654 | AAM12047.1 | |
| AY090482.1 | 21 | 4254 | AAM12048.1 |
More Information
Set Membership: |
|

Genomic Map