Assay Details
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpp | NM_025495.3 | NP_079771.1 | ||
| AK006234.1 | BAB24472.1 | |||
| AK012863.1 | BAB28519.1 | |||
| AK014919.1 | BAB29620.1 | |||
| AK076577.1 | BAC36401.1 | |||
| BC147207.1 | ||||
| BC147208.1 |
Target Gene Details
Entrez Gene ID: | 18295 |
Gene Name: | osteoglycin |
Gene Aliases: |
3110079A16Rik, OG, OIF, SLRR3A, mimecan, mimican |
Location: |
Chr.13:49608071-49624500 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogn | NM_008760.4 | 7 | 1975 | NP_032786.1 |
| AK014259.1 | 1 | 199 | ||
| AK028300.1 | 7 | 2002 | ||
| AK053650.1 | 7 | 1976 | BAC35462.1 | |
| AK160760.1 | 7 | 1997 | BAE35995.1 | |
| AK163772.1 | 6 | 1927 |
More Information
Set Membership: |
|

Genomic Map