Assay Details
Target Gene Details
Entrez Gene ID: | 380839 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 1c |
Gene Aliases: |
EIC, ovalbumin |
Location: |
Chr.13:32881397-32898140 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb1c | NM_173051.2 | 7 | 1333 | NP_766639.2 |
| BC104332.2 | AAI04333.1 | |||
| CJ203237.1 | 1 | 129 |
More Information
Set Membership: |
|

Genomic Map