Assay Details
Target Gene Details
Entrez Gene ID: | 11847 |
Gene Name: | arginase type II |
Gene Aliases: |
AII, AU022422 |
Location: |
Chr.12:79130788-79156301 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arg2 | NM_009705.3 | 8 | 1018 | NP_033835.1 |
| AF032466.1 | 8 | 970 | AAB86959.1 | |
| AK137142.1 | 1 | 2274 | ||
| BC023349.1 | 8 | 982 | AAH23349.1 | |
| U90886.1 | 8 | 990 | AAC22548.1 |
More Information
Set Membership: |
|

Genomic Map