Assay Details
Target Gene Details
Entrez Gene ID: | 26931 |
Gene Name: | protein phosphatase 2, regulatory subunit B', gamma |
Gene Aliases: |
2610043M05Rik, 2700063L20Rik, AI060890, AW545884, C85228, D12Bwg0916e, mKIAA0044 |
Location: |
Chr.12:110447179-110583075 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r5c | NM_001081457.2 | 13 | 3656 | NP_001074926.1 |
| NM_012023.3 | 14 | 3773 | NP_036153.2 | |
| AK011758.1 | 1 | 431 | ||
| AK034227.1 | 1 | 2900 | ||
| AK040459.1 | 1 | 1319 | ||
| AK192619.1 |
More Information
Set Membership: |
|

Genomic Map