Assay Details
Target Gene Details
Entrez Gene ID: | 16364 |
Gene Name: | interferon regulatory factor 4 |
Gene Aliases: |
AI385587, IRF-4, LSIRF, Spip |
Location: |
Chr.13:30749208-30766982 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf4 | NM_013674.1 | 7 | 1112 | NP_038702.1 |
| AK089319.1 | 7 | 1131 | BAC40840.1 | |
| AK089819.1 | 7 | 1134 | BAC40969.1 | |
| AK154138.1 | 7 | 1136 | ||
| AK154315.1 | 7 | 1124 | BAE32509.1 | |
| AK156562.1 | 7 | 1134 | ||
| BC137713.1 | 7 | 1100 | ||
| BC137714.1 | 7 | 1100 | ||
| U11692.1 | 7 | 1112 | AAA75309.1 | |
| U34307.1 | 7 | 1106 | AAA75283.1 |
More Information
Set Membership: |
|

Genomic Map