Assay Details
Target Gene Details
Entrez Gene ID: | 27062 |
Gene Name: | Ca2+-dependent secretion activator |
Gene Aliases: |
AU067781, CAPS, CAPS1, mKIAA1121 |
Location: |
Chr.14:12372560-12823314 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps | NM_001042617.1 | 31 | 4648 | NP_001036082.1 |
| NM_012061.3 | 29 | 4666 | NP_036191.2 | |
| AK122453.1 | 31 | 4648 | BAC65735.1 | |
| AK161919.1 | 19 | 2233 | BAE36633.1 | |
| BC057065.1 | 17 | 2009 | AAH57065.1 | |
| D86214.1 | 29 | 4709 | BAA13044.1 |
More Information
Set Membership: |
|

Genomic Map