Assay Details
Target Gene Details
Entrez Gene ID: | 211948 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, E430028B21Rik |
Location: |
Chr.14:26664117-26669846 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde12 | NM_178668.3 | 1 | 1339 | NP_848783.3 |
| AK082947.1 | 1 | 1162 | BAC38703.1 | |
| AK088843.1 | 1 | 1152 | BAC40606.1 | |
| AK143567.1 | 1 | 1162 | ||
| AK161083.1 | 1 | 1379 | BAE36189.1 | |
| AK167708.1 | 1 | 1137 | BAE39752.1 | |
| BC064450.1 | 1 | 1339 | AAH64450.1 |
More Information
Set Membership: |
|

Genomic Map