Assay Details
Target Gene Details
Entrez Gene ID: | 13665 |
Gene Name: | eukaryotic translation initiation factor 2, subunit 1 alpha |
Gene Aliases: |
0910001O23Rik, 2410026C18Rik, 35kDa, Eif2a, eIF2alpha |
Location: |
Chr.12:78862072-78887010 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2s1 | NM_026114.3 | 8 | 2036 | NP_080390.1 |
| AK009934.1 | 4 | 1437 | BAB26593.1 | |
| AK143499.1 | 8 | 2037 | BAE25400.1 |
More Information
Set Membership: |
|

Genomic Map