Assay Details
Target Gene Details
Entrez Gene ID: | 432713 |
Gene Name: | predicted gene 5441 |
Gene Aliases: |
- |
Location: |
Chr.12:117234521-117337012 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5441 | NR_044987.1 | |||
| AK039060.1 | 2 | 305 |
More Information
Set Membership: |
|

Genomic Map