Assay Details
Target Gene Details
Entrez Gene ID: | 30795 |
Gene Name: | FK506 binding protein 3 |
Gene Aliases: |
25kDa, FKBP-3, FKBP25 |
Location: |
Chr.12:65062432-65073938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp3 | NM_013902.4 | NP_038930.1 | ||
| AF110812.1 | AAD20598.1 | |||
| AF135595.1 | AAD25097.1 | |||
| AK012346.1 | ||||
| AK145648.1 | BAE26564.1 | |||
| AK150802.1 | BAE29865.1 | |||
| AK168641.1 | BAE40500.1 | |||
| BC002122.1 | AAH02122.1 | |||
| U62545.1 | AAB05769.1 |
Target Gene Details
Entrez Gene ID: | 328110 |
Gene Name: | PRP39 pre-mRNA processing factor 39 homolog (yeast) |
Gene Aliases: |
Srcs1 |
Location: |
Chr.12:65036309-65063386 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf39 | NM_177806.3 | 14 | 3414 | NP_808474.2 |
| BC029153.1 | 9 | 2386 | AAH29153.1 |
More Information
Set Membership: |
|

Genomic Map