Assay Details
Target Gene Details
Entrez Gene ID: | 13592 |
Gene Name: | early B cell factor 2 |
Gene Aliases: |
D14Ggc1e, EBF-2, Mmot1, O/E-3, OE-3 |
Location: |
Chr.14:67233292-67430952 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf2 | NM_001276387.1 | 1 | 547 | NP_001263316.1 |
| NM_010095.6 | 1 | 547 | NP_034225.1 | |
| AK053013.1 | 1 | 548 | BAC35239.1 | |
| AK139262.1 | 1 | 254 | BAE23936.1 | |
| AK144416.1 | 1 | 237 |
More Information
Set Membership: |
|

Genomic Map