Assay Details
Target Gene Details
Entrez Gene ID: | 68177 |
Gene Name: | emopamil binding protein-like |
Gene Aliases: |
5730442K12Rik, EBRP |
Location: |
Chr.14:61339763-61360445 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebpl | NM_026598.3 | 4 | 1263 | NP_080874.2 |
| AK044618.1 | 2 | 1751 |
More Information
Set Membership: |
|

Genomic Map