Assay Details
Target Gene Details
Entrez Gene ID: | 238871 |
Gene Name: | phosphodiesterase 4D, cAMP specific |
Gene Aliases: |
9630011N22Rik, Dpde3 |
Location: |
Chr.13:108654177-109955969 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde4d | NM_011056.3 | 17 | 2273 | NP_035186.1 |
| AF536978.1 | 17 | 2273 | AAN10120.1 | |
| AK081466.1 | 10 | 1646 | BAC38226.1 | |
| AY388962.1 | 15 | 1886 | AAQ90406.1 | |
| BC132107.1 | 16 | 2141 | ||
| BC132109.1 | 16 | 2141 | ||
| DQ665891.1 | 10 | 1743 | ||
| DQ665892.1 | 10 | 1529 | ||
| DQ665893.1 | 11 | 1657 | ||
| DQ665894.1 | 11 | 1501 | ||
| DQ665895.1 | 10 | 1784 | ||
| DQ665896.1 | 10 | 1573 | ||
| EU489880.1 | 15 | 2129 |
More Information
Set Membership: |
|

Genomic Map