Assay Details
Target Gene Details
Entrez Gene ID: | 218461 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
B230331L10Rik, C030047E14Rik |
Location: |
Chr.13:95024105-95250397 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde8b | NM_001170669.1 | 19 | 3069 | NP_001164140.1 |
| NM_172263.2 | 20 | 2802 | NP_758467.2 | |
| AK045991.1 | 19 | 3071 | ||
| AK053128.1 | 1 | 1828 | ||
| AK083166.1 | 19 | 2707 | BAC38791.1 | |
| AK138301.1 | 9 | 1736 | BAE23616.1 | |
| AK170192.1 | 5 | 1169 | ||
| BC032286.1 | 20 | 2814 | AAH32286.1 |
More Information
Set Membership: |
|

Genomic Map