Assay Details
Target Gene Details
Entrez Gene ID: | 110794 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), epsilon |
Gene Aliases: |
C/EBPe, CRP1, Gm294 |
Location: |
Chr.14:54710363-54712174 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpe | NM_207131.1 | 2 | 818 | NP_997014.1 |
| AK143561.1 | 2 | 819 | BAE25437.1 | |
| AY570294.1 | 2 | 818 | AAS78198.1 | |
| BC108953.1 | 2 | 755 | AAI08954.1 | |
| BC108954.1 | 2 | 755 | AAI08955.1 |
More Information
Set Membership: |
|

Genomic Map