Assay Details
Target Gene Details
Entrez Gene ID: | 53314 |
Gene Name: | basic leucine zipper transcription factor, ATF-like |
Gene Aliases: |
B-ATF, SFA-2 |
Location: |
Chr.12:85686720-85709087 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Batf | NM_016767.2 | 1 | 91 | NP_058047.1 |
| AF017021.1 | 1 | 91 | AAB70251.1 | |
| AK018587.1 | 1 | 134 | BAB31294.1 |
More Information
Set Membership: |
|

Genomic Map