Assay Details
Target Gene Details
Entrez Gene ID: | 18676 |
Gene Name: | PHD finger protein 2 |
Gene Aliases: |
GRC5 |
Location: |
Chr.13:48801750-48870885 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf2 | NM_011078.3 | 22 | 4293 | NP_035208.2 |
| AK134358.1 | 8 | 2121 | BAE22113.1 | |
| AK172995.1 | 20 | 4000 | BAD32273.1 | |
| BC051633.1 | 22 | 4291 | AAH51633.1 |

Genomic Map