Assay Details
Target Gene Details
Entrez Gene ID: | 217874 |
Gene Name: | retinal degeneration 3-like |
Gene Aliases: |
A930011N13 |
Location: |
Chr.12:111979191-111981128 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rd3l | NM_001127685.1 | 3 | 625 | NP_001121157.1 |
| AK080710.1 | 2 | 1274 | ||
| BC147034.1 | 3 | 625 | ||
| BC147035.1 | 3 | 625 | ||
| BC147747.1 | 3 | 625 | ||
| BC147749.1 | 3 | 625 |
Target Gene Details
Entrez Gene ID: | 74691 |
Gene Name: | tudor domain containing 9 |
Gene Aliases: |
4930441E05Rik |
Location: |
Chr.12:111971474-112068854 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdrd9 | NM_029056.1 | NP_083332.1 | ||
| AB362563.1 |
More Information
Set Membership: |
|

Genomic Map