Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptb | NM_013675.3 | 26 | 6097 | NP_038703.3 |
| AK147299.1 | 26 | 6098 | BAE27831.1 | |
| AK147684.1 | 26 | 5690 | BAE28073.1 | |
| AK147704.1 | 26 | 5690 | BAE28085.1 | |
| AK220551.1 | 7 | 1197 | BAD90537.1 | |
| BC117711.2 | 9 | 1351 | ||
| BC129806.1 | 25 | 5430 | ||
| BC150783.1 | 25 | 5463 | ||
| S66283.1 | 26 | 5573 | AAB28600.1 |

Genomic Map