Assay Details
Target Gene Details
Entrez Gene ID: | 210106 |
Gene Name: | PAP associated domain containing 7 |
Gene Aliases: |
LAK-1, POLK, Pols, TRF4, TRF4-1 |
Location: |
Chr.13:69497952-69534236 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papd7 | NM_001169131.1 | 13 | 3280 | NP_001162602.1 |
| NM_198600.2 | 13 | 3283 | NP_941002.2 | |
| AK152550.1 | 13 | 2981 | BAE31304.1 | |
| BC059846.1 | 13 | 3258 | AAH59846.1 |
More Information
Set Membership: |
|

Genomic Map