Assay Details
Target Gene Details
Entrez Gene ID: | 66761 |
Gene Name: | RIKEN cDNA 4933417A18 gene |
Gene Aliases: |
- |
Location: |
Chr.13:34924405-34956218 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933417A18Rik | NM_025750.3 | 1 | 105 | NP_080026.2 |
| AK016778.1 | 1 | 71 | BAB30424.1 | |
| AK016838.1 | 1 | 106 | BAB30456.1 | |
| BC100450.1 | 1 | 95 | AAI00451.1 | |
| BC145747.1 | 1 | 60 |

Genomic Map