Assay Details
Target Gene Details
Entrez Gene ID: | 12033 |
Gene Name: | B cell receptor associated protein 29 |
Gene Aliases: |
AW208404, Bap29 |
Location: |
Chr.12:31595354-31634658 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcap29 | NM_001164090.1 | 8 | 1451 | NP_001157562.1 |
| NM_007530.3 | 7 | 1614 | NP_031556.1 | |
| AK090068.1 | 8 | 1428 | BAC41076.1 | |
| AK144754.1 | 8 | 1425 | BAE26049.1 | |
| AK205917.1 | 1 | 54 | ||
| AK216433.1 | ||||
| BC003303.1 | 7 | 1518 | AAH03303.1 | |
| BC021661.1 | 7 | 1625 | AAH21661.1 | |
| X78684.1 | 7 | 1426 | CAA55351.1 |
More Information
Set Membership: |
|

Genomic Map