Assay Details
Target Gene Details
Entrez Gene ID: | 58208 |
Gene Name: | B cell leukemia/lymphoma 11B |
Gene Aliases: |
9130430L19Rik, AI604821, B630002E05Rik, BCL-11B, Ctip2, Rit1 |
Location: |
Chr.12:107910403-108004139 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl11b | NM_001079883.1 | 4 | 5588 | NP_001073352.1 |
| NM_001286343.1 | 2 | 5006 | NP_001273272.1 | |
| NM_021399.2 | 3 | 5372 | NP_067374.2 |
More Information
Set Membership: |
|

Genomic Map