Assay Details
Target Gene Details
Entrez Gene ID: | 77974 |
Gene Name: | retinol dehydrogenase 12 |
Gene Aliases: |
A930033N07Rik |
Location: |
Chr.12:79208914-79222661 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh12 | NM_030017.3 | 7 | 1447 | NP_084293.1 |
| AK039233.1 | 7 | 1448 | BAC30288.1 | |
| BC016204.1 | 7 | 1394 | AAH16204.1 |
More Information
Set Membership: |
|

Genomic Map