Assay Details
Target Gene Details
Entrez Gene ID: | 211978 |
Gene Name: | zinc finger, FYVE domain containing 26 |
Gene Aliases: |
4930465A13, 9330197E15Rik, A630028O16Rik, Gm893, mKIAA0321 |
Location: |
Chr.12:79232346-79296310 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfyve26 | NM_001008550.1 | 21 | 4017 | NP_001008550.1 |
| AK135518.1 | ||||
| AK220333.1 | 21 | 3988 | BAD90401.1 | |
| BC141359.1 | 21 | 4000 |

Genomic Map