Assay Details
Target Gene Details
Entrez Gene ID: | 77592 |
Gene Name: | RIKEN cDNA 4931406H21 gene |
Gene Aliases: |
- |
Location: |
Chr.14:25586804-25590661 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931406H21Rik | NR_033492.1 | 1 | 2577 | |
| AK031340.1 | 1 | 2578 | BAC27355.1 | |
| BC147211.1 | 1 | 2178 | ||
| BC147212.1 | 1 | 2178 |
Target Gene Details
Entrez Gene ID: | 328365 |
Gene Name: | zinc finger, MIZ-type containing 1 |
Gene Aliases: |
BC065120, E330020C23, ENSMUSG00000072684, Gm10397, I920194n01, Rai17, Zimp10 |
Location: |
Chr.14:25458865-25666747 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmiz1 | NM_183208.3 | NP_899031.2 | ||
| AK054366.1 | BAC35753.1 | |||
| BC058646.1 | AAH58646.1 | |||
| BC065120.1 | AAH65120.1 |
More Information
Set Membership: |
|

Genomic Map