Assay Details
Target Gene Details
Entrez Gene ID: | 74156 |
Gene Name: | acyl-CoA thioesterase 12 |
Gene Aliases: |
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1 |
Location: |
Chr.13:91741511-91786148 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot12 | NM_028790.3 | 15 | 2533 | NP_083066.1 |
| AK004905.1 | 15 | 2534 | BAB23658.1 | |
| AK034622.1 | 15 | 2537 | BAC28775.1 | |
| BC025852.1 | 15 | 2486 | AAH25852.1 |
More Information
Set Membership: |
|

Genomic Map