Assay Details
Target Gene Details
Entrez Gene ID: | 105014 |
Gene Name: | retinol dehydrogenase 14 (all-trans and 9-cis) |
Gene Aliases: |
3110030G19Rik, AW215439, PAN2 |
Location: |
Chr.12:10390690-10395562 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh14 | NM_023697.2 | 1 | 223 | NP_076186.1 |
| AF303831.1 | 1 | 238 | AAG30904.1 | |
| AK014097.1 | 1 | 192 | ||
| BC020094.1 | 1 | 211 | AAH20094.1 | |
| BC092299.1 | 1 | 219 | AAH92299.1 |
More Information
Set Membership: |
|

Genomic Map