Assay Details
Target Gene Details
Entrez Gene ID: | 210808 |
Gene Name: | laccase (multicopper oxidoreductase) domain containing 1 |
Gene Aliases: |
9030625A04Rik |
Location: |
Chr.14:77024201-77036898 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lacc1 | NM_172488.2 | 3 | 405 | NP_766076.1 |
| AK033565.1 | 3 | 406 | BAC28363.1 | |
| AK053693.1 | 3 | 690 | BAC35476.1 | |
| AK152154.1 | 3 | 346 | BAE30989.1 | |
| AK153349.1 | 3 | 430 | BAE31925.1 | |
| BC116748.1 | 2 | 161 | ||
| BC119140.1 | 2 | 161 |

Genomic Map