Assay Details
Target Gene Details
Entrez Gene ID: | 271127 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 16 |
Gene Aliases: |
- |
Location: |
Chr.13:70727798-70841848 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts16 | NM_172053.2 | 1 | 210 | NP_742050.2 |
| AK031314.1 | 1 | 176 | ||
| AK134523.1 | 1 | 211 | ||
| AK173338.1 | 1 | 146 | BAD32616.1 | |
| BC034739.1 | 1 | 223 | AAH34739.1 |
More Information
Set Membership: |
|

Genomic Map