Assay Details
Target Gene Details
Entrez Gene ID: | 56835 |
Gene Name: | cathepsin R |
Gene Aliases: |
AI428350 |
Location: |
Chr.13:61159215-61164188 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsr | NM_020284.1 | 8 | 1099 | NP_064680.1 |
| AF245399.1 | 8 | 1099 | AAF90051.1 | |
| AK005429.1 | 8 | 1119 | BAB24023.1 | |
| AK014432.1 | 8 | 1115 | BAB29345.1 | |
| AY014778.1 | 8 | 1058 | AAK00507.1 | |
| AY943668.1 | 8 | 1171 | AAX39778.1 | |
| BC100338.1 | 8 | 1094 | AAI00339.1 |
More Information
Set Membership: |
|

Genomic Map