Assay Details
Target Gene Details
Entrez Gene ID: | 17933 |
Gene Name: | myelin transcription factor 1-like |
Gene Aliases: |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Location: |
Chr.12:29528311-29923216 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myt1l | NM_001093775.1 | 25 | 5288 | NP_001087244.1 |
| NM_001093776.1 | 25 | 5194 | NP_001087245.1 | |
| NM_001093778.1 | 25 | 5279 | NP_001087247.1 | |
| NM_008666.3 | 25 | 5282 | NP_032692.2 | |
| AB212898.1 | BAE48255.1 | |||
| AK013847.1 | 1 | 413 | ||
| BC094438.1 | 21 | 4519 |
More Information
Set Membership: |
|

Genomic Map