Assay Details
Target Gene Details
Entrez Gene ID: | 108089 |
Gene Name: | ring finger protein 144A |
Gene Aliases: |
Rnf144, UIP4, Ubce7ip4 |
Location: |
Chr.12:26306794-26415321 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf144a | NM_001081977.2 | 10 | 1847 | NP_001075446.1 |
| NM_080563.4 | 9 | 1773 | NP_542130.1 | |
| AK038586.1 | 9 | 1665 | BAC30058.1 | |
| AK133544.1 | 7 | 1387 | BAE21715.1 | |
| AK172904.1 | 8 | 1608 | BAD32182.1 |
More Information
Set Membership: |
|

Genomic Map