Assay Details
Target Gene Details
Entrez Gene ID: | 104884 |
Gene Name: | tyrosyl-DNA phosphodiesterase 1 |
Gene Aliases: |
2810481F14Rik, 4921509N21Rik, AI838772, AW493413, E430034L06Rik, MLZ-501, SCAN1 |
Location: |
Chr.12:99884515-99955219 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdp1 | NM_028354.4 | 2 | 513 | NP_082630.2 |
| AK013427.1 | ||||
| AK014855.1 | 2 | 494 | ||
| AK033868.1 | 2 | 511 | BAC28500.1 | |
| BC065162.1 | 1 | 483 | AAH65162.1 |
More Information
Set Membership: |
|

Genomic Map