Assay Details
Target Gene Details
Entrez Gene ID: | 26385 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
Gprk6 |
Location: |
Chr.13:55445072-55460927 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grk6 | NM_001038018.4 | 16 | 2050 | NP_001033107.1 |
| NM_001112711.2 | 16 | 2048 | NP_001106182.1 | |
| NM_001286063.1 | 16 | 2030 | NP_001272992.1 | |
| NM_001286064.1 | 16 | 2035 | NP_001272993.1 | |
| NM_001286065.1 | 16 | 1946 | NP_001272994.1 | |
| AF040747.1 | 16 | 1918 | AAC09268.1 | |
| AF040748.1 | 16 | 1920 | AAC09269.1 | |
| AK051405.1 | 16 | 2032 | ||
| AK146119.1 | 16 | 1821 | BAE26914.1 | |
| AK161142.1 | 14 | 1682 | BAE36212.1 | |
| BC075719.1 | 16 | 1931 | AAH75719.1 | |
| Y15798.1 | 16 | 1894 | CAA75789.1 |
More Information
Set Membership: |
|

Genomic Map