Assay Details
Target Gene Details
Entrez Gene ID: | 23894 |
Gene Name: | general transcription factor II H, polypeptide 2 |
Gene Aliases: |
44kDa, BTF2 p44, BTF2-p44, Btf2p44, p44 |
Location: |
Chr.13:100459786-100492610 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2h2 | NM_022011.4 | 16 | 1404 | NP_071294.3 |
| AB221674.1 | ||||
| AK010543.1 | 14 | 1181 | ||
| AK154739.1 | 15 | 1726 | ||
| BC016231.1 | AAH16231.1 | |||
| BC053382.1 | 16 | 1368 | AAH53382.1 | |
| BY480509.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map