Assay Details
Target Gene Details
Entrez Gene ID: | 72852 |
Gene Name: | metallo-beta-lactamase domain containing 2 |
Gene Aliases: |
2900024O10Rik |
Location: |
Chr.13:81711417-81753275 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mblac2 | NM_028372.1 | 2 | 3332 | NP_082648.1 |
More Information
Set Membership: |
|

Genomic Map