Assay Details
Target Gene Details
Entrez Gene ID: | 218121 |
Gene Name: | membrane bound O-acyltransferase domain containing 1 |
Gene Aliases: |
9130215M02Rik, BC023845, LPEAT1, Moact1, Oact1 |
Location: |
Chr.13:30136449-30246694 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat1 | NM_153546.4 | 1 | 178 | NP_705774.1 |
| AB297382.1 | ||||
| AB305045.1 | ||||
| AK020281.1 | 1 | 123 | ||
| AK033388.1 | 1 | 120 | BAC28261.1 | |
| AK137755.1 | 1 | 110 | ||
| AK146984.1 | 1 | 85 | BAE27587.1 | |
| BC023845.1 | 1 | 182 | AAH23845.1 | |
| BC024653.1 | 1 | 130 | AAH24653.1 |
More Information
Set Membership: |
|

Genomic Map