Assay Details
Target Gene Details
Entrez Gene ID: | 13030 |
Gene Name: | cathepsin B |
Gene Aliases: |
CB |
Location: |
Chr.14:63122462-63145923 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsb | NM_007798.3 | 10 | 2895 | NP_031824.1 |
| AK151790.1 | 10 | 2842 | BAE30691.1 | |
| S69034.1 | 10 | 2891 | AAB20536.1 |
More Information
Set Membership: |
|

Genomic Map