Assay Details
Target Gene Details
Entrez Gene ID: | 72935 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 41 |
Gene Aliases: |
2900024F02Rik, AA958953, ABS, AI324246 |
Location: |
Chr.13:55530410-55536658 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx41 | NM_134059.2 | 17 | 2050 | NP_598820.2 |
| AK151427.1 | 17 | 1924 | BAE30391.1 | |
| AK156957.1 | 17 | 1918 | BAE33914.1 | |
| AK168043.1 | 17 | 1923 | BAE40025.1 | |
| BC011308.1 | 17 | 1904 | AAH11308.1 | |
| CJ303880.1 | 2 | 221 |

Genomic Map