Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | 29 | 7922 | NP_001074922.1 |

Genomic Map