Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | 15 | 2088 | NP_001074922.1 |
| NM_001286079.2 | 16 | 2270 | NP_001273008.1 | |
| NM_001286080.2 | 17 | 2317 | NP_001273009.1 | |
| NM_008697.4 | 17 | 2317 | NP_032723.2 | |
| NR_104397.2 | 17 | 2317 | ||
| NR_104398.2 | 16 | 2282 | ||
| AK129394.1 | 17 | 2207 | BAC98204.1 | |
| AK134860.1 | 16 | 2283 | ||
| AK141891.1 | 16 | 2258 | BAE24872.1 | |
| AK162150.1 | 17 | 2261 | ||
| AK167264.1 | 14 | 3304 | ||
| AY515727.1 | 15 | 2067 | AAS87211.1 | |
| BC137789.1 | 16 | 2155 | ||
| BC144913.1 | 17 | 2202 | ||
| U40342.1 | 17 | 2230 | AAA83234.1 |

Genomic Map