Assay Details
Target Gene Details
Entrez Gene ID: | 18080 |
Gene Name: | ninein |
Gene Aliases: |
3110068G20Rik, AI385615, AU024711, mKIAA1565 |
Location: |
Chr.12:70011435-70113830 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nin | NM_001081453.1 | NP_001074922.1 | ||
| NM_001286079.2 | 29 | 6423 | NP_001273008.1 | |
| NM_001286080.2 | 30 | 6443 | NP_001273009.1 | |
| NM_008697.4 | 30 | 6470 | NP_032723.2 | |
| NR_104397.2 | 31 | 6531 | ||
| NR_104398.2 | 29 | 4375 | ||
| AK129394.1 | 30 | 6360 | BAC98204.1 | |
| AK134860.1 | 29 | 4375 | ||
| AK147187.1 | 13 | 3226 | ||
| AK167264.1 | 26 | 5309 | ||
| AY515727.1 | AAS87211.1 | |||
| BC137789.1 | 29 | 6308 | ||
| BC144913.1 | 30 | 6355 | ||
| CF586279.1 | 1 | 366 | ||
| U40342.1 | 30 | 6350 | AAA83234.1 |

Genomic Map